N-Allyl-2-pyrrolidinone 95%

Supplier: Apollo Scientific

Synonyms: 1-Allylpyrrolidin-2-one

OR906382-1G OR906382-5G
APOSOR906382-1GEA 112 GBP
APOSOR906382-1G APOSOR906382-5G
N-Allyl-2-pyrrolidinone 95%
N-Allyl-2-pyrrolidinone
Formula: C₇H₁₁NO
MW: 125,168 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00152226
CAS Number: 2687-97-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR