2-(3-Chlorobenzyl)pyrrolidine 96%

Supplier: Apollo Scientific

OR906298-1G OR906298-250MG
APOSOR906298-1GEA 394 GBP
APOSOR906298-1G APOSOR906298-250MG
2-(3-Chlorobenzyl)pyrrolidine 96%
2-(3-Chlorobenzyl)pyrrolidine
Formula: C₁₁H₁₄ClN
MW: 195,689 g/mol
Storage Temperature: Ambient
MDL Number: MFCD02663447
CAS Number: 383127-29-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR