Ethyl (2s)-2-(1H-pyrrol-1-yl)propanoate 95%
Supplier: Apollo Scientific
OR905579-1G
OR905579-250MG
APOSOR905579-1GEA
780
GBP
APOSOR905579-1G
APOSOR905579-250MG
Ethyl (2s)-2-(1H-pyrrol-1-yl)propanoate 95%
Ethyl (2s)-2-(1H-pyrrol-1-yl)propanoate
Formula:
C₉H₁₃NO₂ MW: 167,205 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD08752770 CAS Number: 935765-03-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...