1-(2,5-Dimethylphenyl)-1H-pyrrole 95%
Supplier: Apollo Scientific
OR905161-1G
OR905161-250MG
APOSOR905161-1GEA
780
GBP
APOSOR905161-1G
APOSOR905161-250MG
1-(2,5-Dimethylphenyl)-1H-pyrrole 95%
1-(2,5-Dimethylphenyl)-1H-pyrrole
Formula:
C₁₂H₁₃N MW: 171,238 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD02665222 CAS Number: 37560-44-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...