2-(2-Formyl-1H-pyrrol-1-yl)benzonitrile 95%
Supplier: Apollo Scientific
OR904650-1G
OR904650-250MG
APOSOR904650-1GEA
780
GBP
APOSOR904650-1G
APOSOR904650-250MG
2-(2-Formyl-1H-pyrrol-1-yl)benzonitrile 95%
2-(2-Formyl-1H-pyrrol-1-yl)benzonitrile
Formula:
C₁₂H₈N₂O MW: 196,205 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD02664851 CAS Number: 68557-19-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...