2-(4-Isopropylphenyl)pyrrolidine 95%
Supplier: Apollo Scientific
OR904501-1G
OR904501-250MG
APOSOR904501-1GEA
728
GBP
APOSOR904501-1G
APOSOR904501-250MG
2-(4-Isopropylphenyl)pyrrolidine 95%
2-(4-Isopropylphenyl)pyrrolidine
Formula:
C₁₃H₁₉N MW: 189,297 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD05189447 CAS Number: 881040-01-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...