1-Methyl-2-(2-pyrrolidinyl)piperidine 95%
Supplier: Apollo Scientific
OR904335-1G
OR904335-250MG
APOSOR904335-1GEA
728
GBP
APOSOR904335-1G
APOSOR904335-250MG
1-Methyl-2-(2-pyrrolidinyl)piperidine 95%
1-Methyl-2-(2-pyrrolidinyl)piperidine
Formula:
C₁₀H₂₀N₂ MW: 168,279 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD02663424 CAS Number: 383127-17-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...