1-(3,5-Dimethylphenyl)-1H-pyrrole 95%
Supplier: Apollo Scientific
OR904244-1G
OR904244-250MG
APOSOR904244-1GEA
780
GBP
APOSOR904244-1G
APOSOR904244-250MG
1-(3,5-Dimethylphenyl)-1H-pyrrole 95%
1-(3,5-Dimethylphenyl)-1H-pyrrole
Formula:
C₁₂H₁₃N MW: 171,238 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD02665241 CAS Number: 37560-45-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...