1-(2-Isopropylphenyl)-1H-pyrrole 95%
Supplier: Apollo Scientific
OR903579-1G
OR903579-250MG
APOSOR903579-1GEA
780
GBP
APOSOR903579-1G
APOSOR903579-250MG
1-(2-Isopropylphenyl)-1H-pyrrole 95%
1-(2-Isopropylphenyl)-1H-pyrrole
Formula:
C₁₃H₁₅N MW: 185,265 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD02665233 CAS Number: 77503-16-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...