(2-Pyrrolidin-1-ylphenyl)methanol 95%

Supplier: Apollo Scientific

OR902654-1G OR902654-5G
APOSOR902654-1GEA 197 GBP
APOSOR902654-1G APOSOR902654-5G
(2-Pyrrolidin-1-ylphenyl)methanol 95%
(2-Pyrrolidin-1-ylphenyl)methanol
Formula: C₁₁H₁₅NO
MW: 177,243 g/mol
Storage Temperature: Ambient
MDL Number: MFCD08690257
CAS Number: 73051-88-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR