2-(3,5-Dimethylphenyl)pyrrolidine 95%

Supplier: Apollo Scientific

OR902508-1G OR902508-250MG
APOSOR902508-1GEA 728 GBP
APOSOR902508-1G APOSOR902508-250MG
2-(3,5-Dimethylphenyl)pyrrolidine 95%
2-(3,5-Dimethylphenyl)pyrrolidine
Formula: C₁₂H₁₇N
MW: 175,27 g/mol
Storage Temperature: Ambient
MDL Number: MFCD02663469
CAS Number: 383127-44-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR