1-(3-Methyl-pyridin-2-yl)-1H-pyrrole-2-carbaldehyde 95%

Supplier: Apollo Scientific

OR902467-1G OR902467-250MG
APOSOR902467-1GEA 453 GBP
APOSOR902467-1G APOSOR902467-250MG
1-(3-Methyl-pyridin-2-yl)-1H-pyrrole-2-carbaldehyde 95%
1-(3-Methyl-pyridin-2-yl)-1H-pyrrole-2-carbaldehyde
Formula: C₁₁H₁₀N₂O
MW: 186,21 g/mol
Storage Temperature: Ambient
MDL Number: MFCD02665158
CAS Number: 383136-11-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR