2-tert-Butoxy-p-picoline 98%

Supplier: Apollo Scientific

OR901890-1G OR901890-5G
APOSOR901890-1GEA 132 GBP
APOSOR901890-1G APOSOR901890-5G
2-tert-Butoxy-p-picoline 98%
2-tert-Butoxy-p-picoline
Formula: C₁₀H₁₅NO
MW: 165,232 g/mol
Storage Temperature: Ambient
MDL Number: MFCD18914537
CAS Number: 57883-15-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR