5-Bromo-2-methylnicotinonitrile 95%

Supplier: Apollo Scientific

OR901872-1G OR901872-250MG
APOSOR901872-1GEA 392 GBP
APOSOR901872-1G APOSOR901872-250MG
5-Bromo-2-methylnicotinonitrile 95%
5-Bromo-2-methylnicotinonitrile
Formula: C₇H₅BrN₂
MW: 197,032 g/mol
Storage Temperature: Ambient
MDL Number: MFCD18255108
CAS Number: 956276-47-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR