N-Acetyl-D,L-homotryptophan

Supplier: Apollo Scientific

Synonyms: a-Acetamido-g-(3-indole)butanoic acid

OR7840T-10MG OR7840T-50MG
APOSOR7840T-10MGEA 205 GBP
APOSOR7840T-10MG APOSOR7840T-50MG
N-Acetyl-D,L-homotryptophan
N-Acetyl-D,L-homotryptophan
Formula: C₁₄H₁₆N₂O₃
Melting Pt: 112…113 °C
Storage Temperature: Ambient
MDL Number: MFCD00189457
CAS Number: 205813-00-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR