5-Chloro-2-iodoanisole 97%

Supplier: Apollo Scientific
Warning

Synonyms: 5-Chloro-2-iodophenyl methyl ether, 4-Chloro-1-iodo-2-methoxybenzene

OR70062-1G OR70062-250MG
APOSOR70062-1GEA 44.6 GBP
APOSOR70062-1G APOSOR70062-250MG
5-Chloro-2-iodoanisole 97%
5-Chloro-2-iodoanisole
Formula: C₇H₆ClIO
MW: 268,479 g/mol
Boiling Pt: 135 °C (10 mm Hg)
Melting Pt: 20 °C
Storage Temperature: Ambient
MDL Number: MFCD09753735
CAS Number: 755027-21-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR