2-Amino-6-bromo-3-chlorophenol

Supplier: Apollo Scientific
Warning

Synonyms: 3-Bromo-6-chloro-2-hydroxyaniline

OR61257-1G OR61257-250MG
APOSOR61257-1GEA 434 GBP
APOSOR61257-1G APOSOR61257-250MG
2-Amino-6-bromo-3-chlorophenol
2-Amino-6-bromo-3-chlorophenol
Formula: C₆H₅BrClNO
MW: 222,467 g/mol
Storage Temperature: Ambient
MDL Number: MFCD20036248
CAS Number: 1332765-75-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR