4-(4-Methylhomopiperazin-1-yl)methylbenzyl alcohol
Supplier: Apollo Scientific
Synonyms:
{4-[(4-Methylperhydro-1,4-diazepin-1-yl)methyl]phenyl}methanol
OR6073-1G
OR6073-5G
APOSOR6073-1GEA
119
GBP
APOSOR6073-1G
APOSOR6073-5G
4-(4-Methylhomopiperazin-1-yl)methylbenzyl alcohol
4-(4-Methylhomopiperazin-1-yl)methylbenzyl alcohol
Formula:
C₁₄H₂₂N₂O MW: 234,337 g/mol Boiling Pt: 190…200 °C (2 mm Hg) Storage Temperature: Ambient |
MDL Number:
MFCD09064973 CAS Number: 884507-50-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...