3,5-Dimethylisonicotinonitrile

Supplier: Apollo Scientific
Danger

Synonyms: 3,5-Dimethylpyridine-4-carbonitrile, 4-Cyano-3,5-dimethylpyridine

OR60147-250MG OR60147-1G
APOSOR60147-250MGEA 296 GBP
APOSOR60147-250MG APOSOR60147-1G
3,5-Dimethylisonicotinonitrile
3,5-Dimethylisonicotinonitrile
Formula: C₈H₈N₂
MW: 132,163 g/mol
Melting Pt: 88…89 °C
Storage Temperature: Refrigerator
MDL Number: MFCD11052634
CAS Number: 7584-08-9
UN: 3439
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR