2-{4-[Methyl(aminomethyl)]phenyl}-5-methyl-1,3,4-oxadiazole 95%

Supplier: Apollo Scientific
Danger

Synonyms: N-Methyl-1-[4-(5-methyl-1,3,4-oxadiazol-2-yl)phenyl]methylamine

OR59897-1G OR59897-250MG
APOSOR59897-1GEA 254 GBP
APOSOR59897-1G APOSOR59897-250MG
2-{4-[Methyl(aminomethyl)]phenyl}-5-methyl-1,3,4-oxadiazole 95%
2-{4-[Methyl(aminomethyl)]phenyl}-5-methyl-1,3,4-oxadiazole
Formula: C₁₁H₁₃N₃O
MW: 203,24 g/mol
Melting Pt: 63…65 °C
Storage Temperature: Ambient
MDL Number: MFCD11109326
CAS Number: 944450-83-3
UN: 3259
ADR: 8,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR