5-(Fur-2-yl)-2-[methyl(aminomethyl)]pyridine 95%

Supplier: Apollo Scientific
Danger

Synonyms: [5-(Fur-2-yl)pyridin-2-yl]-N-methylmethylamine

OR59896-1G OR59896-250MG
APOSOR59896-1GEA 255 GBP
APOSOR59896-1G APOSOR59896-250MG
5-(Fur-2-yl)-2-[methyl(aminomethyl)]pyridine 95%
5-(Fur-2-yl)-2-[methyl(aminomethyl)]pyridine
Formula: C₁₁H₁₂N₂O
Boiling Pt: 110 °C (0,08 mm Hg)
Storage Temperature: Refrigerator
MDL Number: MFCD10700054
CAS Number: 892502-04-4
UN: 2735
ADR: 8,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR