3-Bromo-5-[(4-methoxybenzyl)oxy]pyridine

Supplier: Apollo Scientific

OR59410-1G OR59410-5G
APOSOR59410-1GEA 119 GBP
APOSOR59410-1G APOSOR59410-5G
3-Bromo-5-[(4-methoxybenzyl)oxy]pyridine
3-Bromo-5-[(4-methoxybenzyl)oxy]pyridine
Formula: C₁₃H₁₂BrNO₂
MW: 294,144 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09878436
CAS Number: 552331-73-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR