2,6-Bis(dimethylamino)pyrimidine-4-carboxylic acid monohydrate 95+%

Supplier: Apollo Scientific
Warning

Synonyms: 2,6-Bis(dimethylamino)-1,3-diazine-4-carboxylic acid monohydrate, 2,4-Bis(dimethylamino)pyrimidine-6-carboxylic acid monohydrate

OR5664-1G OR5664-250MG
APOSOR5664-1GEA 43.4 GBP
APOSOR5664-1G APOSOR5664-250MG
2,6-Bis(dimethylamino)pyrimidine-4-carboxylic acid monohydrate 95+%
2,6-Bis(dimethylamino)pyrimidine-4-carboxylic acid monohydrate
Formula: C₉H₁₄N₄O₂·H₂O
MW: 228,248 g/mol
Melting Pt: 164…168 °C
Storage Temperature: Ambient
MDL Number: MFCD07366474
CAS Number: 1052402-84-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR