Ethyl 6-aminonicotinate

Supplier: Apollo Scientific
Warning

Synonyms: Ethyl 2-aminopyridine-5-carboxylate

OR5577-1G OR5577-5G
APOSOR5577-1GEA 51.2 GBP
APOSOR5577-1G APOSOR5577-5G
Ethyl 6-aminonicotinate
Ethyl 6-aminonicotinate
Formula: C₈H₁₀N₂O₂
MW: 166,177 g/mol
Melting Pt: 145…147 °C
Storage Temperature: Refrigerator
MDL Number: MFCD04972544
CAS Number: 39658-41-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR