5-Bromo-1-chloro-2-methoxy-3-nitrobenzene 95%
Supplier: Apollo Scientific
|
Warning
|
OR55555-1G
OR55555-250MG
APOSOR55555-1GEA
591
GBP
APOSOR55555-1G
APOSOR55555-250MG
5-Bromo-1-chloro-2-methoxy-3-nitrobenzene 95%
5-Bromo-1-chloro-2-methoxy-3-nitrobenzene
Formula:
C₇H₅BrClNO₃ MW: 266,477 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD31653056 CAS Number: 2091142-75-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...