Methyl 2-iodo-3-methylbenzoate 98%

Supplier: Apollo Scientific
Warning

OR55476-1G OR55476-5G
APOSOR55476-1GEA 95.7 GBP
APOSOR55476-1G APOSOR55476-5G
Methyl 2-iodo-3-methylbenzoate 98%
Methyl 2-iodo-3-methylbenzoate
Formula: C₉H₉IO₂
MW: 276,071 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD12923249
CAS Number: 103441-60-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR