(1-Methyl-1,2,4-triazol-3-yl)methanamine 90%

Supplier: Apollo Scientific
Warning

OR55400-250MG OR55400-1G
APOSOR55400-250MGEA 381 GBP
APOSOR55400-250MG APOSOR55400-1G
(1-Methyl-1,2,4-triazol-3-yl)methanamine 90%
(1-Methyl-1,2,4-triazol-3-yl)methanamine
Formula: C₄H₈N₄
MW: 112,133 g/mol
Storage Temperature: Ambient
MDL Number: MFCD17255308
CAS Number: 785760-73-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR