(1-Bromopropyl)benzene
Supplier: Apollo Scientific
|
Warning
|
Synonyms:
1-Bromopropylbenzene
OR55292-1G
OR55292-250MG
APOSOR55292-1GEA
774
GBP
APOSOR55292-1G
APOSOR55292-250MG
(1-Bromopropyl)benzene
(1-Bromopropyl)benzene
Formula:
C₉H₁₁Br MW: 199,088 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00055549 CAS Number: 2114-36-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...