2-Chloro-4-(methylthio)pyridine

Supplier: Apollo Scientific
Warning

OR55260-250MG OR55260-1G
APOSOR55260-250MGEA 157 GBP
APOSOR55260-250MG APOSOR55260-1G
2-Chloro-4-(methylthio)pyridine
2-Chloro-4-(methylthio)pyridine
Formula: C₆H₆ClNS
MW: 159.637 g/mol
Storage Temperature: Ambient
MDL Number: MFCD07437890
CAS Number: 71506-83-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR