5-(4-Bromophenyl)pyrimidine-4,6-diol

Supplier: Apollo Scientific
Warning

Synonyms: 5-(4-Bromophenyl)-6-hydroxypyrimidin-4(1H)-one

OR55029-10G OR55029-1G OR55029-5G
APOSOR55029-10GEA 87.8 GBP
APOSOR55029-10G APOSOR55029-1G APOSOR55029-5G
5-(4-Bromophenyl)pyrimidine-4,6-diol
5-(4-Bromophenyl)pyrimidine-4,6-diol
Formula: C₁₀H₇BrN₂O₂
MW: 267,079 g/mol
Melting Pt: >300 °C
Storage Temperature: Ambient
MDL Number: MFCD13151898
CAS Number: 706811-25-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR