5-Methylpyrimidine-2-carboxylic acid
Supplier: Apollo Scientific
|
Warning
|
OR54460-1G
OR54460-250MG
APOSOR54460-1GEA
174
GBP
APOSOR54460-1G
APOSOR54460-250MG
5-Methylpyrimidine-2-carboxylic acid
5-Methylpyrimidine-2-carboxylic acid
Formula:
C₆H₆N₂O₂ MW: 138,124 g/mol Boiling Pt: 362,8 °C Melting Pt: 157…161 °C Flash Pt: 173 °C Storage Temperature: Ambient |
MDL Number:
MFCD09910349 CAS Number: 99420-75-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...