2-Amino-5-chloro-N,3-dimethylbenzamide 95%

Supplier: Apollo Scientific

OR52935-100MG OR52935-1G OR52935-250MG
APOSOR52935-100MGEA 191 GBP
APOSOR52935-100MG APOSOR52935-1G APOSOR52935-250MG
2-Amino-5-chloro-N,3-dimethylbenzamide 95%
2-Amino-5-chloro-N,3-dimethylbenzamide
Formula: C₉H₁₁ClN₂O
MW: 198,649 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09030601
CAS Number: 890707-28-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR