3-Bromo-2-nitrobenzyl alcohol 95%

Supplier: Apollo Scientific
Warning

Synonyms: (3-Bromo-2-nitrophenyl)methanol

OR52700-1G OR52700-250MG OR52700-5G
APOSOR52700-1GEA 262 GBP
APOSOR52700-1G APOSOR52700-250MG APOSOR52700-5G
3-Bromo-2-nitrobenzyl alcohol 95%
3-Bromo-2-nitrobenzyl alcohol
Formula: C₇H₆BrNO₃
MW: 232,031 g/mol
Storage Temperature: Ambient
MDL Number: MFCD18392284
CAS Number: 1261475-45-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR