4-Bromo-7-chloro-6-(4-cyclopropylbenzyl)-2,3-dihydrobenzofuran 95%
Supplier: Apollo Scientific
|
Warning
|
OR52693-100MG
OR52693-50MG
OR52693-1G
APOSOR52693-100MGEA
591
GBP
APOSOR52693-100MG
APOSOR52693-50MG
APOSOR52693-1G
4-Bromo-7-chloro-6-(4-cyclopropylbenzyl)-2,3-dihydrobenzofuran 95%
4-Bromo-7-chloro-6-(4-cyclopropylbenzyl)-2,3-dihydrobenzofuran
Formula:
C₁₈H₁₆BrClO MW: 363,676 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD29919372 CAS Number: 1415477-23-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...