Methyl (2-hydroxyphenyl)acetate

Supplier: Apollo Scientific
Warning

Synonyms: Methyl 2-(2-hydroxyphenyl)acetate

OR52466-10G OR52466-1G OR52466-5G
APOSOR52466-10GEA 106 GBP
APOSOR52466-10G APOSOR52466-1G APOSOR52466-5G
Methyl (2-hydroxyphenyl)acetate
Methyl (2-hydroxyphenyl)acetate
Formula: C₉H₁₀O₃
MW: 166,174 g/mol
Melting Pt: 71…72 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00090077
CAS Number: 22446-37-3
EINECS: 245-004-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR