Methyl 4-amino-2,5-dimethylbenzoate 95+%

Supplier: Apollo Scientific
Warning

OR52228-250MG OR52228-1G OR52228-5G
APOSOR52228-250MGEA 64.3 GBP
APOSOR52228-250MG APOSOR52228-1G APOSOR52228-5G
Methyl 4-amino-2,5-dimethylbenzoate 95+%
Methyl 4-amino-2,5-dimethylbenzoate
Formula: C₁₀H₁₃NO₂
MW: 179,216 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09839220
CAS Number: 21339-74-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR