Phenyl(piperidin-4-yl)methanol hydrochloride

Supplier: Apollo Scientific
Warning

OR52222-1G OR52222-5G
APOSOR52222-1GEA 165 GBP
APOSOR52222-1G APOSOR52222-5G
Phenyl(piperidin-4-yl)methanol hydrochloride
Phenyl(piperidin-4-yl)methanol hydrochloride
Formula: C₁₂H₁₇NO·ClH
MW: 227,73 g/mol
Storage Temperature: Ambient
MDL Number: MFCD28014028
CAS Number: 91688-34-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR