2-(Tetrahydro-2H-pyran-4-yloxy)benzaldehyde

Supplier: Apollo Scientific
Warning

OR5203-1G OR5203-5G
APOSOR5203-1GEA 257 GBP
APOSOR5203-1G APOSOR5203-5G
2-(Tetrahydro-2H-pyran-4-yloxy)benzaldehyde
2-(Tetrahydro-2H-pyran-4-yloxy)benzaldehyde
Formula: C₁₂H₁₄O₃
MW: 206,238 g/mol
Melting Pt: 67…70 °C
Storage Temperature: Ambient
MDL Number: MFCD09064958
CAS Number: 898289-31-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR