4-Chlorobenzo[c][1,2,5]thiadiazole 95+%

Supplier: Apollo Scientific
Warning

OR500034-1G OR500034-250MG
APOSOR500034-1GEA 430 GBP
APOSOR500034-1G APOSOR500034-250MG
4-Chlorobenzo[c][1,2,5]thiadiazole 95+%
4-Chlorobenzo[c][1,2,5]thiadiazole
Formula: C₆H₃ClN₂S
MW: 170,619 g/mol
Storage Temperature: Ambient
MDL Number: MFCD17168971
CAS Number: 2207-28-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR