4-(5-Bromo-1-benzofuran-2-yl)pyrimidin-2-amine

Supplier: Apollo Scientific
Warning

OR48104-1G OR48104-250MG
APOSOR48104-1GEA 188 GBP
APOSOR48104-1G APOSOR48104-250MG
4-(5-Bromo-1-benzofuran-2-yl)pyrimidin-2-amine
4-(5-Bromo-1-benzofuran-2-yl)pyrimidin-2-amine
Formula: C₁₂H₈BrN₃O
Storage Temperature: Ambient
MDL Number: MFCD07381956
CAS Number: 1275259-26-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR