1-Bromo-3-cyclopropyl-5-nitrobenzene

Supplier: Apollo Scientific

OR48083-1G OR48083-250MG
APOSOR48083-1GEA 519 GBP
APOSOR48083-1G APOSOR48083-250MG
1-Bromo-3-cyclopropyl-5-nitrobenzene
1-Bromo-3-cyclopropyl-5-nitrobenzene
Formula: C₉H₈BrNO₂
MW: 242,069 g/mol
Storage Temperature: Freezer
MDL Number: MFCD32640245
CAS Number: 1211541-80-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR