2-Bromomethyl-1,3,5-trimethylbenzene 95%

Supplier: Apollo Scientific

Synonyms: 2-(Bromomethyl)-1,3,5-trimethylbenzene

OR480803-1G OR480803-5G
APOSOR480803-1GEA 83.8 GBP
APOSOR480803-1G APOSOR480803-5G
2-Bromomethyl-1,3,5-trimethylbenzene 95%
2-Bromomethyl-1,3,5-trimethylbenzene
Formula: C₁₀H₁₃Br
Storage Temperature: Ambient
MDL Number: MFCD09056829
CAS Number: 4761-00-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR