2,4,6-Trichloro-3-nitropyridine 95%
Supplier: Apollo Scientific
OR480779-1G
OR480779-5G
APOSOR480779-1GEA
141
GBP
APOSOR480779-1G
APOSOR480779-5G
2,4,6-Trichloro-3-nitropyridine 95%
2,4,6-Trichloro-3-nitropyridine
Formula:
C₅HCl₃N₂O₂ MW: 227,433 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD09264535 CAS Number: 60186-13-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...