2-(Methylthio)-4-thien-2-ylpyrimidine ≥95%
Supplier: Apollo Scientific
OR480658-25G
OR480658-5G
APOSOR480658-25GEA
611
GBP
APOSOR480658-25G
APOSOR480658-5G
2-(Methylthio)-4-thien-2-ylpyrimidine ≥95%
2-(Methylthio)-4-thien-2-ylpyrimidine
Formula:
C₉H₈N₂S₂ MW: 208,303 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00206706 CAS Number: 683274-58-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...