1-(4-Bromophenyl)-3-ethoxypropan-1-one 97%

Supplier: Apollo Scientific

OR480606-1G OR480606-5G
APOSOR480606-1GEA 195 GBP
APOSOR480606-1G APOSOR480606-5G
1-(4-Bromophenyl)-3-ethoxypropan-1-one 97%
1-(4-Bromophenyl)-3-ethoxypropan-1-one
Formula: C₁₁H₁₃BrO₂
MW: 257,124 g/mol
Storage Temperature: Ambient
MDL Number: MFCD12546969
CAS Number: 859954-71-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR