(1S)-1-(3-Bromophenyl)-N-methylethanamine hydrochloride
Supplier: Apollo Scientific
|
Warning
|
OR475073-1G
OR475073-250MG
APOSOR475073-1GEA
207
GBP
APOSOR475073-1G
APOSOR475073-250MG
(1S)-1-(3-Bromophenyl)-N-methylethanamine hydrochloride
(1S)-1-(3-Bromophenyl)-N-methylethanamine hydrochloride
Formula:
C₉H₁₂BrN·ClH MW: 250,563 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD31695803 CAS Number: 2391975-25-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...