Ethyl 2-(4-oxocyclohexyl)butanoate 95%

Supplier: Apollo Scientific

OR471711-1G OR471711-250MG
APOSOR471711-1GEA 715 GBP
APOSOR471711-1G APOSOR471711-250MG
Ethyl 2-(4-oxocyclohexyl)butanoate 95%
Ethyl 2-(4-oxocyclohexyl)butanoate
Formula: C₁₂H₂₀O₃
MW: 212,285 g/mol
Storage Temperature: Ambient
MDL Number: MFCD31695762
CAS Number: 1923824-52-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR