2,5-Dimethoxypyridine 95%
Supplier: Apollo Scientific
OR471688-10G
OR471688-1G
OR471688-5G
APOSOR471688-10GEA
362
GBP
APOSOR471688-10G
APOSOR471688-1G
APOSOR471688-5G
2,5-Dimethoxypyridine 95%
2,5-Dimethoxypyridine
Formula:
C₇H₉NO₂ MW: 139,152 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD12546620 CAS Number: 867267-24-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...