5-Nitro-2,3-dihydro-1,3-benzoxazol-2-one 95%

Supplier: Apollo Scientific

OR471670-10G OR471670-1G OR471670-5G
APOSOR471670-10GEA 879 GBP
APOSOR471670-10G APOSOR471670-1G APOSOR471670-5G
5-Nitro-2,3-dihydro-1,3-benzoxazol-2-one 95%
5-Nitro-2,3-dihydro-1,3-benzoxazol-2-one
Formula: C₇H₄N₂O₄
MW: 180,118 g/mol
Storage Temperature: Ambient
MDL Number: MFCD01929029
CAS Number: 3889-13-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR