4-Isopropylsalicylaldehyde

Supplier: Apollo Scientific

Synonyms: 2-Hydroxy-4-isopropylbenzaldehyde

OR471497-1G OR471497-5G
APOSOR471497-1GEA 94.4 GBP
APOSOR471497-1G APOSOR471497-5G
4-Isopropylsalicylaldehyde
4-Isopropylsalicylaldehyde
Formula: C₁₀H₁₂O₂
MW: 164,201 g/mol
Storage Temperature: Ambient
MDL Number: MFCD16997144
CAS Number: 536-32-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR